Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024381.2 | NP_077357.2 | 17, 18 | 1586 | ||
XM_006256987.2 | XP_006257049.1 | 18, 19 | 1638 | |||
XM_006256988.2 | XP_006257050.1 | 17, 18 | 1620 | |||
XM_008773274.1 | XP_008771496.1 | 19, 20 | 1703 | |||
XM_008773275.1 | XP_008771497.1 | 18, 19 | 1685 | |||
XM_008773276.1 | XP_008771498.1 | 19, 20 | 1703 | |||
XM_008773278.1 | XP_008771500.1 | 19, 20 | 1702 | |||
GenBank mRNA | CO557097.1 | - | 2, 3 | 268 | ||
CO564445.1 | - | 8, 9 | 719 | |||
D16102.1 | - | 17, 18 | 1589 |