Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mef2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030860.2 | NP_110487.2 | 8 | 1376 | ||
XM_006232771.1 | XP_006232833.1 | 8 | 975 | |||
XM_006232772.1 | XP_006232834.1 | 8 | 972 | |||
XM_006232773.1 | XP_006232835.1 | 7 | 951 | |||
GenBank mRNA | AJ005425.1 | - | 7 | 951 | ||
BC081790.1 | - | 8 | 1376 | 7134848 |