Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh6a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031057.2 | NP_112319.2 | 2, 3 | 151 | ||
XM_006240354.2 | XP_006240416.1 | 2, 3 | 516 | |||
XM_008764863.1 | XP_008763085.1 | 2, 3 | 153 | |||
GenBank mRNA | CB760062.1 | - | 2, 3 | 208 | ||
DN948946.1 | - | 2, 3 | 148 |