Gene Symbol: Pax4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031799.1 | NP_113987.1 | 5 | 564 | ||
XM_006236231.2 | XP_006236293.1 | 6 | 6617 | |||
XM_006236232.2 | XP_006236294.1 | 7 | 1613 | |||
XM_006236233.2 | XP_006236295.1 | 7 | 1055 | |||
GenBank mRNA | AF053100.1 | - | 5 | 564 | ||
AF053102.1 | - | 5 | 564 | |||
AF198155.1 | - | 4 | 516 | |||
AF198156.1 | - | 4 | 516 |