Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nae1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC127481.1 | - | 13 | 1033 | 8370668 | |
FM118995.1 | - | 7 | 599 | |||
FM131178.1 | - | 4 | 269 | |||
FQ234218.1 | - | 2 | 128 | |||
U90829.1 | - | 13 | 1030 |