Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gjb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053984.2 | NP_446436.1 | 1 | 870 | ||
XM_008763985.1 | XP_008762207.1 | 2 | 600 | |||
XM_008763986.1 | XP_008762208.1 | 1 | 198 | |||
GenBank mRNA | X76168.1 | - | 1 | 870 |