Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3gat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054003.1 | NP_446455.1 | 5 | 1131 | ||
XM_006242733.2 | XP_006242795.1 | 5 | 1403 | |||
XM_006242734.2 | XP_006242796.1 | 5 | 1392 | |||
GenBank mRNA | D88035.1 | - | 5 | 1131 |