Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133398.1 | NP_596889.1 | 6 | 1331 | ||
XM_006241520.2 | XP_006241582.1 | 6 | 2750 | |||
XM_006241521.2 | XP_006241583.1 | 5 | 2685 | |||
XM_006241523.2 | XP_006241585.1 | 5 | 2682 | |||
XM_006241524.2 | XP_006241586.1 | 6 | 2748 | |||
XM_006241525.2 | XP_006241587.1 | 5 | 2683 | |||
GenBank mRNA | AF100421.2 | - | 6 | 1331 | ||
FQ225234.1 | - | 3 | 413 | |||
FQ225997.1 | - | 3 | 413 | |||
FQ226094.1 | - | 6 | 1327 | |||
FQ227915.1 | - | 4 | 479 | |||
FQ231703.1 | - | 3 | 413 | |||
FQ233989.1 | - | 1 | 177 | |||
FQ234381.1 | - | 2 | 264 | |||
FQ234551.1 | - | 5 | 1269 |