Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adora2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053294.4 | NP_445746.3 | 3 | 854 | ||
GenBank mRNA | AF115508.1 | - | 2 | 360 | ||
AF228684.1 | - | 2 | 460 | |||
BC081727.1 | - | 3 | 854 | 7121208 | ||
L08102.1 | - | 2 | 426 | |||
M91214.2 | - | 2 | 711 | |||
S47609.1 | - | 2 | 500 |
Set Membership: |
G-protein coupled receptor |