Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl8a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173110.1 | NP_775133.1 | 6 | 613 | ||
XM_006253907.1 | XP_006253969.1 | 6 | 623 | |||
XM_006253908.1 | XP_006253970.1 | 6 | 579 | |||
GenBank mRNA | BC097274.1 | - | 5 | 560 | 7441946 | |
BC158577.1 | - | 6 | 607 | 7461869 | ||
M76537.1 | - | 6 | 590 | |||
U09099.1 | - | 6 | 613 |