Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: l7Rn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013897.1 | NP_001013919.1 | 4 | 589 | ||
XM_006229662.2 | XP_006229724.1 | 3 | 556 | |||
XM_008759638.1 | XP_008757860.1 | 3 | 387 | |||
XM_008759639.1 | XP_008757861.1 | 2 | 316 | |||
GenBank mRNA | BC088340.1 | - | 4 | 589 | 7367791 | |
FQ212130.1 | - | 3 | 500 | |||
FQ213824.1 | - | 3 | 501 | |||
FQ214677.1 | - | 3 | 501 | |||
FQ215506.1 | - | 3 | 492 | |||
FQ216197.1 | - | 3 | 501 | |||
FQ216668.1 | - | 3 | 501 | |||
FQ216759.1 | - | 3 | 501 | |||
FQ218863.1 | - | 3 | 498 | |||
FQ220044.1 | - | 3 | 501 | |||
FQ220310.1 | - | 3 | 552 | |||
FQ220575.1 | - | 3 | 501 | |||
FQ224272.1 | - | 3 | 503 | |||
FQ224945.1 | - | 3 | 501 | |||
FQ229506.1 | - | 3 | 497 |