Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1303003 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004225.1 | NP_001004225.1 | 5 | 460 | ||
GenBank mRNA | BC079380.1 | - | 5 | 460 | 7110981 | |
FQ213331.1 | - | 5 | 470 | |||
FQ217777.1 | - | 5 | 476 | |||
FQ231770.1 | - | 5 | 457 |