Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1303003 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004225.1 | NP_001004225.1 | 3 | 265 | ||
GenBank mRNA | BC079380.1 | - | 3 | 265 | 7110981 | |
FQ213331.1 | - | 3 | 275 | |||
FQ217777.1 | - | 3 | 281 | |||
FQ231770.1 | - | 3 | 262 |