Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gjb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017251.2 | NP_058947.1 | 2 | 234 | ||
XM_006257061.2 | XP_006257123.1 | 3 | 386 | |||
XM_008773255.1 | XP_008771477.1 | 2 | 1117 | |||
GenBank mRNA | BC078868.1 | - | 2 | 198 | 7127952 | |
CK476084.1 | - | 2 | 234 | |||
FQ210131.1 | - | 2 | 219 | |||
X04070.1 | - | 1 | 180 |