Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017275.1 | NP_058971.1 | 3, 4 | 405 | ||
XM_006229572.2 | XP_006229634.1 | 3, 4 | 237 | |||
XM_006229573.2 | XP_006229635.1 | 3, 4 | 261 | |||
XM_006229574.2 | XP_006229636.1 | 3, 4 | 211 | |||
XM_006229575.2 | XP_006229637.1 | 3, 4 | 215 | |||
XM_008773628.1 | XP_008771850.1 | 3, 4 | 330 | |||
XM_008773629.1 | XP_008771851.1 | 2, 3 | 208 | |||
GenBank mRNA | AB004267.1 | - | 3, 4 | 405 | ||
D86556.1 | - | 3, 4 | 256 |