Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox7a2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106704.1 | NP_001100174.1 | 3 | 546 | ||
XM_006239680.1 | XP_006239742.1 | 3 | 375 | |||
GenBank mRNA | BC166896.1 | - | 3 | 358 | 6921504 | |
FQ212738.1 | - | 3 | 324 | |||
FQ213890.1 | - | 3 | 358 | |||
FQ215773.1 | - | 3 | 327 | |||
FQ220707.1 | - | 3 | 358 | |||
FQ230066.1 | - | 3 | 358 |