Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1563072 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107981.1 | NP_001101451.1 | 2 | 312 | ||
XM_006238868.1 | XP_006238930.1 | 2 | 254 | |||
XM_006238869.2 | XP_006238931.1 | 2 | 869 | |||
XM_006238870.2 | XP_006238932.1 | 3 | 905 |
Gene Symbol: LOC100912566 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750034.3 | XP_003750082.1 | 869 | |||
XM_006238915.1 | XP_006238977.1 | 254 | ||||
XM_006238916.1 | XP_006238978.1 | 183 | ||||
XM_006238917.2 | XP_006238979.1 | 905 |