Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prrg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108482.1 | NP_001101952.1 | 3 | 323 | ||
XM_006229098.2 | XP_006229160.1 | 3 | 981 | |||
XM_008759396.1 | XP_008757618.1 | 4 | 411 | |||
XM_008759397.1 | XP_008757619.1 | 3 | 272 |