Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NKX2-2 SIRNA Assays ›
Gene Symbol:
Nkx2-2
Gene Name:
NK2 homeobox 2
Gene Aliases:
-
Chromosome Location:
Chr.3: 141408798 - 141411170 on Build Rnor_6.0
UniGene ID:
Rn.32651
Species:
Rat
siRNA ID
RSS343066
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Nkx2-2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001191904.1
NP_001178833.1
2
634
Back To Top
More Information
Panther Classification:
Molecular Function -
homeodomain transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
type B pancreatic cell development
pancreatic A cell fate commitment
type B pancreatic cell fate commitment
pancreatic PP cell fate commitment
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
smoothened signaling pathway
nervous system development
response to glucose
positive regulation of gene expression
oligodendrocyte development
response to organic cyclic compound
spinal cord motor neuron differentiation
spinal cord oligodendrocyte cell differentiation
spinal cord oligodendrocyte cell fate specification
optic nerve development
cell differentiation
endocrine pancreas development
response to progesterone
regulation of cell differentiation
positive regulation of cell differentiation
negative regulation of neuron differentiation
positive regulation of neuron differentiation
positive regulation of transcription from RNA polymerase II promoter
cell development
digestive tract development
neuron fate specification
astrocyte differentiation
positive regulation of oligodendrocyte differentiation
positive regulation of sequence-specific DNA binding transcription factor activity
ventral spinal cord interneuron fate determination
RNA polymerase II distal enhancer sequence-specific DNA binding
core promoter proximal region DNA binding
transcriptional activator activity, RNA polymerase II distal enhancer sequence-specific binding
molecular_function
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
transcription factor binding
sequence-specific DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-xql29:80/10.239.165.153:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0