Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RUNX1 SIRNA Assays ›
Gene Symbol:
Runx1
Gene Name:
runt-related transcription factor 1
Gene Aliases:
Aml1, Cbfa2
Chromosome Location:
Chr.11: 32765147 - 32860628 on Build Rnor_6.0
UniGene ID:
Rn.11201
Species:
Rat
siRNA ID
RSS343419
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Runx1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_017325.1
NP_059021.1
3, 4
937
XM_008768598.1
XP_008766820.1
3, 4
2739
GenBank mRNA
L35271.1
-
3, 4
937
Back To Top
More Information
Panther Classification:
Molecular Function -
Runt transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
skeletal system development
in utero embryonic development
liver development
myeloid progenitor cell differentiation
transcription, DNA-templated
regulation of transcription, DNA-templated
central nervous system development
negative regulation of cell proliferation
regulation of signal transduction
hemopoiesis
myeloid cell differentiation
neuron differentiation
negative regulation of granulocyte differentiation
positive regulation of granulocyte differentiation
hair follicle morphogenesis
response to retinoic acid
positive regulation of interleukin-2 production
embryonic hemopoiesis
positive regulation of angiogenesis
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
behavioral response to pain
neuron development
definitive hemopoiesis
regulation of hair follicle cell proliferation
cellular response to transforming growth factor beta stimulus
positive regulation of cell maturation
positive regulation of progesterone secretion
regulatory region DNA binding
RNA polymerase II regulatory region sequence-specific DNA binding
core promoter binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
molecular_function
DNA binding
transcription factor activity, sequence-specific DNA binding
calcium ion binding
ATP binding
transcription factor binding
protein homodimerization activity
protein heterodimerization activity
repressing transcription factor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-wh4bk:80/10.239.163.60:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0