Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tagln3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035236.1 | NP_001030313.1 | 2, 3 | 332 | ||
NM_031676.2 | NP_113864.2 | 1, 2 | 280 | |||
GenBank mRNA | AF459788.1 | - | 2, 3 | 476 | ||
CB795348.1 | - | 1, 2 | 321 | |||
FQ212703.1 | - | 1, 2 | 274 | |||
FQ212858.1 | - | 1, 2 | 274 | |||
FQ214377.1 | - | 1, 2 | 273 | |||
FQ214483.1 | - | 1, 2 | 274 | |||
FQ214564.1 | - | 1, 2 | 274 | |||
M84725.1 | - | 1, 2 | 254 |
Gene Symbol: LOC103693564 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768676.1 | XP_008766898.1 | 2, 3 | 549 |