Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnl3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081958.1 | NP_001075427.1 | 11, 12 | 1084 | ||
GenBank mRNA | CB783942.1 | - | 1, 2 | 123 | ||
FQ223839.1 | - | 722 |