Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100125364 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103356.2 | NP_001096826.1 | 2 | 343 | ||
XM_006227969.2 | XP_006228031.1 | 2 | 276 | |||
XM_006227970.2 | XP_006228032.1 | 2 | 254 | |||
XM_008758746.1 | XP_008756968.1 | 2 | 349 | |||
GenBank mRNA | BC079175.1 | - | 2 | 343 | 7130104 |