Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133296.1 | NP_579830.1 | 5, 6 | 825 | ||
GenBank mRNA | S76742.1 | - | 5, 6 | 825 |