Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mpp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053668.1 | NP_446120.1 | 3 | 216 | ||
XM_008767931.1 | XP_008766153.1 | 3 | 192 | |||
XM_008767932.1 | XP_008766154.1 | 3 | 316 | |||
GenBank mRNA | AF087697.1 | - | 1 | 68 | ||
FQ211573.1 | - | 3 | 342 |