Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Lim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053771.1 | NP_446223.1 | 4 | 405 | ||
GenBank mRNA | L04191.1 | - | 4 | 383 | ||
M87053.1 | - | 4 | 405 | |||
S55224.1 | - | 4 | 396 |