Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacng4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080692.1 | NP_542423.1 | 4 | 938 | ||
GenBank mRNA | AF361341.1 | - | 4 | 938 |