Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138832.1 | NP_620187.1 | 4, 5 | 542 | ||
XM_006242277.2 | XP_006242339.1 | 4, 5 | 601 | |||
XM_006242278.2 | XP_006242340.1 | 5, 6 | 810 | |||
XM_006242279.2 | XP_006242341.1 | 5, 6 | 780 | |||
XM_006242280.2 | XP_006242342.1 | 4, 5 | 649 | |||
XM_006242281.2 | XP_006242343.1 | 4, 5 | 690 | |||
GenBank mRNA | AF075704.1 | - | 4, 5 | 542 | ||
BC097283.1 | - | 4, 5 | 462 | 7444889 |