Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dkc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133419.1 | NP_596910.1 | 10 | 1094 | ||
XM_008773596.1 | XP_008771818.1 | 10 | 1540 | |||
GenBank mRNA | BC087600.1 | - | 10 | 1053 | ||
BC099832.1 | - | 10 | 1052 | 7934021 | ||
Z34922.1 | - | 10 | 1094 |