Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uggt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133596.1 | NP_598280.1 | 39 | 4351 | ||
XM_006244734.2 | XP_006244796.1 | 39 | 4573 | |||
XM_006244735.2 | XP_006244797.1 | 38 | 4549 | |||
GenBank mRNA | AF200359.1 | - | 39 | 4351 |