Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cblb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133601.1 | NP_598285.1 | 11, 12 | 1642 | ||
XM_006248258.2 | XP_006248320.1 | 11, 12 | 1716 | |||
XM_006248259.2 | XP_006248321.1 | 11, 12 | 1799 | |||
XM_008768626.1 | XP_008766848.1 | 10, 11 | 1617 | |||
XM_008768627.1 | XP_008766849.1 | 11, 12 | 1912 | |||
GenBank mRNA | AB071283.1 | - | 11, 12 | 1642 |