Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: A3galt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138524.2 | NP_612533.1 | 4, 5 | 410 | ||
GenBank mRNA | AF248543.1 | - | 4, 5 | 409 | ||
BC061714.1 | - | 4, 5 | 410 | 5598151 |