Search Thermo Fisher Scientific
Gene Symbol: Ero1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138528.1 | NP_612537.1 | 11 | 785 | ||
GenBank mRNA | AF489855.1 | - | 11 | 772 | ||
AY071924.1 | - | 11 | 785 | |||
AY071925.1 | - | 8 | 467 | |||
FQ228432.1 | - | 630 |