Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pbld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138530.2 | NP_612539.2 | 846 | |||
XM_006256372.2 | XP_006256434.1 | 10 | 1161 | |||
GenBank mRNA | AY083160.1 | - | 858 | |||
BC078735.1 | - | 846 | 7126368 | |||
FQ209640.1 | - | 881 | ||||
FQ219037.1 | - | 881 |
Gene Symbol: Pbld2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002728946.3 | XP_002728992.2 | 12 | 1869 |