Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cela2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012553.2 | NP_036685.1 | 7, 8 | 823 | ||
GenBank mRNA | FQ226773.1 | - | 7, 8 | 825 | ||
FQ227675.1 | - | 7, 8 | 825 | |||
FQ228000.1 | - | 7, 8 | 825 | |||
FQ228239.1 | - | 6, 7 | 706 | |||
V01233.1 | - | 7, 8 | 823 |