Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Epo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017001.1 | NP_058697.1 | 3 | 199 | ||
XM_006249101.2 | XP_006249163.1 | 2 | 341 | |||
GenBank mRNA | D10763.1 | - | 3 | 199 | ||
L10608.1 | - | 2 | 168 |