Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ibsp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012587.2 | NP_036719.2 | 2, 3 | 117 | ||
GenBank mRNA | AB001383.1 | - | 1, 2 | 44 | ||
BC127506.1 | - | 2, 3 | 113 | 8369751 | ||
DQ213013.1 | - | 1, 2 | 44 | |||
DY314698.1 | - | 2, 3 | 117 | |||
J04215.1 | - | 2, 3 | 120 |