Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082477.2 | NP_001075946.2 | 3 | 299 | ||
NM_001082478.1 | NP_001075947.1 | 3 | 596 | |||
NM_001082479.1 | NP_001075948.1 | 3 | 596 | |||
NM_178866.4 | NP_849197.3 | 3 | 299 | |||
XM_006241191.2 | XP_006241253.1 | 4 | 440 | |||
XM_006241192.1 | XP_006241254.1 | 4 | 354 | |||
XM_006241193.2 | XP_006241255.1 | 4 | 439 | |||
XM_006241194.2 | XP_006241256.1 | 3 | 478 | |||
GenBank mRNA | BC086374.1 | - | 3 | 299 | 7300903 | |
D00698.1 | - | 3 | 280 | |||
FQ209824.1 | - | 4 | 428 | |||
FQ210560.1 | - | 3 | 312 | |||
FQ210688.1 | - | 3 | 312 | |||
FQ220385.1 | - | 3 | 620 | |||
FQ225388.1 | - | 3 | 312 | |||
FQ225437.1 | - | 3 | 312 | |||
FQ231951.1 | - | 3 | 312 | |||
FQ234943.1 | - | 3 | 312 | |||
M17335.1 | - | 3 | 496 | |||
X06043.1 | - | 2 | 181 | |||
X06107.1 | - | 3 | 574 | |||
X06108.1 | - | 4 | 388 |