Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139188.3 | NP_631927.1 | 4 | 205 | ||
XM_008767358.1 | XP_008765580.1 | 5 | 813 | |||
XM_008767359.1 | XP_008765581.1 | 4 | 571 | |||
GenBank mRNA | AY062254.1 | - | 4 | 233 |