Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdkn1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033757.1 | NP_001028929.1 | 1 | 464 | ||
NM_001033758.1 | NP_001028930.1 | 1 | 464 | |||
NM_182735.2 | NP_877399.2 | 2 | 329 | |||
XM_008760076.1 | XP_008758298.1 | 2 | 376 | |||
XM_008760077.1 | XP_008758299.1 | 2 | 382 | |||
XM_008760078.1 | XP_008758300.1 | 2 | 336 | |||
XM_008760079.1 | XP_008758301.1 | 2 | 334 | |||
GenBank mRNA | AJ488291.1 | - | 1 | 191 | ||
AY351678.1 | - | 1 | 465 | |||
AY351679.1 | - | 1 | 465 | |||
AY576608.1 | - | 2 | 328 | |||
AY586728.1 | - | 2 | 293 | |||
BC098646.1 | - | 2 | 327 | 7109418 | ||
CB775146.1 | - | 2 | 288 |