Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TNNT3 SIRNA Assays ›
Gene Symbol:
Tnnt3
Gene Name:
troponin T type 3 (skeletal, fast)
Gene Aliases:
Tnt
Chromosome Location:
Chr.1: 215666628 - 215683628 on Build Rnor_6.0
UniGene ID:
Rn.15488
Species:
Rat
siRNA ID
RSS351728
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Tnnt3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001270664.1
NP_001257593.1
12
591
NM_001270665.1
NP_001257594.1
14
624
NM_001270666.1
NP_001257595.1
14
624
NM_001270668.1
NP_001257597.1
12
591
NM_001270670.1
NP_001257599.1
13
606
NM_001270671.1
NP_001257600.1
13
606
NM_001270672.1
NP_001257601.1
12
597
NM_001270673.1
NP_001257602.1
13
609
NM_001270675.1
NP_001257604.1
12
597
NM_001270676.1
NP_001257605.1
13
609
NM_001270677.1
NP_001257606.1
11
579
NM_001270678.1
NP_001257607.1
13
612
NM_031532.2
NP_113720.1
13
612
XM_006230681.2
XP_006230743.1
12
554
XM_006230683.2
XP_006230745.1
11
542
XM_006230685.2
XP_006230747.1
10
524
XM_006230686.2
XP_006230748.1
10
524
XM_008760080.1
XP_008758302.1
15
658
XM_008760081.1
XP_008758303.1
15
658
XM_008760082.1
XP_008758304.1
14
608
XM_008760083.1
XP_008758305.1
14
605
XM_008760084.1
XP_008758306.1
13
631
XM_008760085.1
XP_008758307.1
13
596
XM_008760086.1
XP_008758308.1
12
578
GenBank mRNA
DQ062204.1
-
13
508
DQ273678.1
-
13
508
FQ214650.1
-
13
572
FQ214722.1
-
12
559
FQ214766.1
-
12
553
FQ214833.1
-
13
574
FQ215030.1
-
12
553
FQ215059.1
-
12
584
FQ215106.1
-
12
553
FQ215114.1
-
12
559
FQ215252.1
-
13
571
FQ215316.1
-
12
573
FQ215327.1
-
14
586
FQ215374.1
-
13
599
FQ215598.1
-
14
583
FQ215602.1
-
12
586
FQ215694.1
-
12
591
FQ215881.1
-
14
586
FQ215885.1
-
14
574
FQ215897.1
-
13
599
FQ215955.1
-
12
553
FQ216024.1
-
9
553
FQ216051.1
-
12
553
FQ216059.1
-
14
581
FQ216097.1
-
13
567
FQ216131.1
-
12
584
FQ216208.1
-
13
571
FQ216324.1
-
12
553
FQ216367.1
-
12
553
FQ216417.1
-
13
572
FQ216467.1
-
12
553
FQ216480.1
-
12
559
FQ216535.1
-
13
568
FQ216561.1
-
9
553
FQ216694.1
-
13
568
FQ216711.1
-
12
590
FQ216750.1
-
9
444
FQ216763.1
-
10
544
FQ216784.1
-
12
559
FQ216866.1
-
14
586
FQ216925.1
-
4
315
FQ217147.1
-
12
552
FQ217587.1
-
2
62
FQ217846.1
-
14
618
FQ217900.1
-
13
599
FQ223840.1
-
12
553
FQ223999.1
-
12
553
FQ224006.1
-
12
555
FQ224345.1
-
4
284
FQ224794.1
-
12
553
FQ224813.1
-
14
586
FQ224836.1
-
12
553
FQ224839.1
-
13
571
FQ224892.1
-
13
574
FQ224966.1
-
13
557
Back To Top
More Information
Panther Classification:
Molecular Function -
actin binding motor protein
Gene Ontology Categories:
Function(s)
Process(es)
skeletal muscle contraction
muscle contraction
regulation of muscle contraction
regulation of striated muscle contraction
metabolic process
regulation of ATPase activity
actin binding
tropomyosin binding
troponin C binding
calcium-dependent ATPase activity
troponin I binding
troponin T binding
calcium-dependent protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-qtpg5:80/10.239.161.70:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline