Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TNNT3 SIRNA Assays ›
Gene Symbol:
Tnnt3
Gene Name:
troponin T type 3 (skeletal, fast)
Gene Aliases:
Tnt
Chromosome Location:
Chr.1: 215666628 - 215683628 on Build Rnor_6.0
UniGene ID:
Rn.15488
Species:
Rat
siRNA ID
RSS351729
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Tnnt3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001270664.1
NP_001257593.1
12
645
NM_001270665.1
NP_001257594.1
14
678
NM_001270666.1
NP_001257595.1
14
678
NM_001270668.1
NP_001257597.1
12
645
NM_001270670.1
NP_001257599.1
13
660
NM_001270671.1
NP_001257600.1
13
660
NM_001270672.1
NP_001257601.1
12
651
NM_001270673.1
NP_001257602.1
13
663
NM_001270675.1
NP_001257604.1
12
651
NM_001270676.1
NP_001257605.1
13
663
NM_001270677.1
NP_001257606.1
11
633
NM_001270678.1
NP_001257607.1
13
666
NM_031532.2
NP_113720.1
13
666
XM_006230681.2
XP_006230743.1
12
608
XM_006230683.2
XP_006230745.1
11
596
XM_006230685.2
XP_006230747.1
10
578
XM_006230686.2
XP_006230748.1
10
578
XM_008760080.1
XP_008758302.1
15
712
XM_008760081.1
XP_008758303.1
15
712
XM_008760082.1
XP_008758304.1
14
662
XM_008760083.1
XP_008758305.1
14
659
XM_008760084.1
XP_008758306.1
13
685
XM_008760085.1
XP_008758307.1
13
650
XM_008760086.1
XP_008758308.1
12
632
GenBank mRNA
DQ062204.1
-
13
562
DQ273678.1
-
13
562
FQ214650.1
-
13
626
FQ214722.1
-
12
613
FQ214766.1
-
12
607
FQ214833.1
-
13
628
FQ215030.1
-
12
607
FQ215059.1
-
12
638
FQ215106.1
-
12
607
FQ215252.1
-
13
625
FQ215316.1
-
12
627
FQ215319.1
-
12
607
FQ215327.1
-
14
640
FQ215374.1
-
13
653
FQ215598.1
-
14
636
FQ215602.1
-
12
640
FQ215694.1
-
12
645
FQ215723.1
-
14
679
FQ215881.1
-
14
640
FQ215885.1
-
14
628
FQ215897.1
-
13
653
FQ215955.1
-
12
607
FQ216024.1
-
9
607
FQ216051.1
-
12
607
FQ216097.1
-
13
621
FQ216131.1
-
12
638
FQ216208.1
-
13
625
FQ216324.1
-
12
607
FQ216367.1
-
12
607
FQ216467.1
-
12
607
FQ216480.1
-
12
613
FQ216535.1
-
13
622
FQ216561.1
-
9
607
FQ216694.1
-
13
622
FQ216711.1
-
12
644
FQ216750.1
-
9
498
FQ216784.1
-
12
613
FQ216866.1
-
14
640
FQ216925.1
-
4
369
FQ217147.1
-
12
606
FQ217587.1
-
2
116
FQ217846.1
-
14
672
FQ217900.1
-
13
653
FQ223840.1
-
12
607
FQ223999.1
-
12
607
FQ224006.1
-
12
609
FQ224794.1
-
12
607
FQ224836.1
-
12
607
FQ224839.1
-
13
625
FQ224892.1
-
13
628
FQ224966.1
-
13
611
Back To Top
More Information
Panther Classification:
Molecular Function -
actin binding motor protein
Gene Ontology Categories:
Function(s)
Process(es)
skeletal muscle contraction
muscle contraction
regulation of muscle contraction
regulation of striated muscle contraction
metabolic process
regulation of ATPase activity
actin binding
tropomyosin binding
troponin C binding
calcium-dependent ATPase activity
troponin I binding
troponin T binding
calcium-dependent protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0