Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fhl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033926.2 | NP_001029098.1 | 2 | 217 | ||
NM_001271199.1 | NP_001258128.1 | 3 | 209 | |||
NM_001271200.1 | NP_001258129.1 | 2 | 150 | |||
NM_001271201.1 | NP_001258130.1 | 2 | 150 | |||
NM_145669.3 | NP_663702.1 | 2 | 217 | |||
XM_006257671.2 | XP_006257733.1 | 2 | 334 | |||
XM_006257672.2 | XP_006257734.1 | 3 | 270 | |||
XM_006257673.2 | XP_006257735.1 | 3 | 293 | |||
XM_006257674.2 | XP_006257736.1 | 3 | 287 | |||
XM_006257675.2 | XP_006257737.1 | 2 | 243 | |||
XM_006257676.2 | XP_006257738.1 | 2 | 213 | |||
XM_008773647.1 | XP_008771869.1 | 2 | 173 | |||
GenBank mRNA | AF134773.1 | - | 3 | 209 | ||
AY115564.1 | - | 2 | 179 | |||
BC061782.1 | - | 2 | 168 | 5623247 | ||
CB702630.1 | - | 2 | 191 | |||
CK477468.1 | - | 1 | 123 | |||
FM042472.1 | - | 2 | 150 | |||
FQ119120.1 | - | 2 | 217 |