Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fhl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033926.2 | NP_001029098.1 | 4 | 475 | ||
NM_001271199.1 | NP_001258128.1 | 5 | 467 | |||
NM_001271200.1 | NP_001258129.1 | 4 | 408 | |||
NM_001271201.1 | NP_001258130.1 | 4 | 408 | |||
NM_145669.3 | NP_663702.1 | 4 | 475 | |||
XM_006257671.2 | XP_006257733.1 | 4 | 592 | |||
XM_006257672.2 | XP_006257734.1 | 5 | 528 | |||
XM_006257673.2 | XP_006257735.1 | 5 | 551 | |||
XM_006257674.2 | XP_006257736.1 | 5 | 545 | |||
XM_006257675.2 | XP_006257737.1 | 4 | 501 | |||
XM_006257676.2 | XP_006257738.1 | 4 | 471 | |||
XM_008773647.1 | XP_008771869.1 | 4 | 431 | |||
GenBank mRNA | AF134773.1 | - | 5 | 467 | ||
AY115564.1 | - | 4 | 437 | |||
BC061782.1 | - | 4 | 426 | 5623247 | ||
CK477468.1 | - | 3 | 381 | |||
FM042472.1 | - | 4 | 408 | |||
FQ119120.1 | - | 4 | 475 |