Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr37l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145784.2 | NP_665727.2 | 2 | 793 | ||
GenBank mRNA | AF087947.1 | - | 859 | |||
BC168215.1 | - | 2 | 793 | 7313732 |
Set Membership: |
![]() |