Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cst3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012837.1 | NP_036969.1 | 1 | 90 | ||
GenBank mRNA | BC087591.1 | - | 1 | 30 | 7307694 | |
CB608544.1 | - | 1 | 73 | |||
CB692474.1 | - | 1 | 100 | |||
FQ210306.1 | - | 1 | 43 | |||
FQ211221.1 | - | 1 | 43 | |||
FQ211526.1 | - | 1 | 43 | |||
FQ211840.1 | - | 1 | 43 | |||
FQ211941.1 | - | 1 | 43 | |||
FQ212057.1 | - | 1 | 72 | |||
FQ212112.1 | - | 1 | 43 | |||
FQ212277.1 | - | 1 | 43 | |||
FQ212367.1 | - | 1 | 43 | |||
FQ212374.1 | - | 1 | 43 | |||
FQ214337.1 | - | 1 | 43 | |||
FQ218053.1 | - | 1 | 74 | |||
FQ221782.1 | - | 1 | 43 | |||
FQ222820.1 | - | 1 | 43 | |||
FQ223224.1 | - | 1 | 43 | |||
FQ224103.1 | - | 1 | 43 | |||
FQ228332.1 | - | 1 | 40 | |||
FQ228399.1 | - | 1 | 43 | |||
FQ234324.1 | - | 1 | 74 |