Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cav1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031556.3 | NP_113744.2 | 2 | 293 | ||
NM_133651.2 | NP_598412.2 | 1 | 285 | |||
XM_006236130.2 | XP_006236192.1 | 2 | 265 | |||
XM_006236131.1 | XP_006236193.1 | 2 | 232 | |||
GenBank mRNA | AF439779.1 | - | 1 | 45 | ||
BC078744.1 | - | 2 | 172 | |||
BC104689.1 | - | 1 | 108 | |||
BC128717.1 | - | 1 | 115 | |||
BC161826.1 | - | 1 | 278 | 9100114 | ||
CB698936.1 | - | 2 | 187 | |||
CK364516.1 | - | 1 | 285 | |||
FM039194.1 | - | 2 | 293 | |||
Z46614.1 | - | 2 | 166 |