Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clrn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153299.2 | NP_695211.2 | 2 | 401 | ||
GenBank mRNA | AF482698.1 | - | 2 | 401 |