Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bles03 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024233.2 | NP_001019404.2 | 2 | 895 | ||
NM_001243534.1 | NP_001230463.1 | 2 | 909 | |||
GenBank mRNA | AF502571.2 | - | 1 | 102 | ||
BC093387.1 | - | 2 | 754 | 7389419 | ||
FQ233132.1 | - | 2 | 788 |