Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vimp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173120.2 | NP_775143.2 | 4 | 378 | ||
GenBank mRNA | AF367467.1 | - | 4 | 388 | ||
BC059111.1 | - | 4 | 382 | 6920258 | ||
FQ210095.1 | - | 4 | 370 | |||
FQ218853.1 | - | 4 | 388 | |||
FQ219944.1 | - | 4 | 370 | |||
FQ220082.1 | - | 4 | 373 | |||
FQ225575.1 | - | 4 | 388 | |||
FQ229642.1 | - | 4 | 370 |